SP Wilmad-LabGlass NMR Tube Caps, PTFE, SP Industries
Supplier: SP Industries
WG-1264-3 WG-1264-4 WG-1264-5 WG-1264-10 WG-1265-100
76459-686PK 64.15 USD
76459-686 76459-688 76459-690 76459-692 76459-694
SP Wilmad-LabGlass NMR Tube Caps, PTFE, SP Industries
Closures Test/Sample Tube Closures
The PTFE NMR tube caps are machined to exact specifications with a smaller gyroradius and more homogeneous mass distribution than disposable caps for better spinning stability.
These caps are recommended in experiments at high to ultra high field and experiments using chloroform as reference solution.
Caution: Only to be used with Wilmad precision NMR tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable