DuPont™ Tyvek® Medical Grade 1073B Autoclave Wrappers, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MGTVKSHEET0203 MGTVKSHEET0303 MGTVKSHEET0306 MGTVKSHEET0404 MGTVKSHEET0406 MGTVKSHEET0518 MGTVKSHEET0536 MGTVKSHEET0606 MGTVKSHEET0707 MGSHEET7.5X7.5 MGTVKSHEET0808 MGTVKSHEET0810 MGTVKSHEET0836 MGTVKSHEET0909 MGTVKSHEET0911 MGTVKSHEET1010 MGTVKSHEET1036 MGTVKSHEET1212 MGTVKSHEET1515 MGTVKSHEET1616 MGTVKSHEET1818 MGTVKSHEET1836 MGTVKSHEET1860 MGTVKSHEET2020 MGTVKSHEET2424 MGTVKSHEET2430 MGTVKSHEET3030 MGTVKSHEET2854 MGTVKSHEET3636 MGTVKSHEET4040 MGTVKSHEET4080 MGTVKSHEET5454 MGTVKSHEET6060 MGTYSHEET60140
76450-752CS 564.86 USD
76450-752 76450-754 76450-756 76450-758 76450-760 76450-762 76450-764 76450-768 76450-770 76450-772 76450-774 76450-776 76450-778 76450-780 76450-782 76450-786 76450-788 76450-790 76450-794 76450-796 76450-798 76450-800 76450-802 76450-804 76450-806 76450-808 76450-810 76450-812 76450-814 76450-816 76450-818 76450-820 76450-822 76450-824
DuPont™ Tyvek® Medical Grade 1073B Autoclave Wrappers, Keystone Cleanroom Products
Sterilization Wraps
Medical grade 1073B Tyvek® wrappers improve the microbial barrier and cleanliness of your sterilization wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Medical grade 1073B Tyvek® wrappers are an improvement over cellulose or SMS based wrapping materials for steam sterilization. Eliminate the wasted time and expense of cutting larger square sheets of sterilization wrap to your desired sizes. Clean cut squares and rectangles are delivered in labeled,More Product Information