Cyto-Chex® BCT Blood Collection Tubes, Streck
Supplier: Streck Laboratories
213362 213361 213559 218980 213386
87005-196BX 600.01 USD
87005-196 87005-282 76449-902 76449-904 89029-967
Cyto-Chex® BCT Blood Collection Tubes, Streck
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Cyto-Chex® BCT is a direct draw blood collection tube that maintains cellular morphology and surface antigen expression, including cluster of differentiation (CD) markers prior to analysis by flow cytometry.
  • Up to 14-day sample stability when analyzed by flow cytometry
  • FDA 510(k) cleared for consistent recovery of HIV-associated lymphocyte subsets for 14 days
  • Available in 2 and 5 ml vials
Whole blood samples collected in Cyto-Chex® BCT can be stored at room temperature for up to 14 days. Cyto-Chex® BCT has been FDA 510(k) cleared for consistentMore Product Information