EZBio® Bioprocess, Single-Use, Double Bagged, Sterile Carboys, Foxx Life Sciences
Supplier: VWR International
76449-290 76449-292 76449-294 76449-296 76449-298 76449-300 76449-302 76449-304 76449-306 76449-308 76449-310 76449-312 76449-314 76449-316 76449-318 76449-320 76449-322 76449-324 76449-326 76449-328 76449-330 76449-332 76449-334 76449-336 76449-338 76449-340 76449-342 76449-344 76449-346 76449-348 76449-350 76449-352 76449-354 76449-356 76449-358 76449-360
76449-290EA 206.02 USD
76449-290 76449-292 76449-294 76449-296 76449-298 76449-300 76449-302 76449-304 76449-306 76449-308 76449-310 76449-312 76449-314 76449-316 76449-318 76449-320 76449-322 76449-324 76449-326 76449-328 76449-330 76449-332 76449-334 76449-336 76449-338 76449-340 76449-342 76449-344 76449-346 76449-348 76449-350 76449-352 76449-354 76449-356 76449-358 76449-360
EZBio® Bioprocess, Single-Use, Double Bagged, Sterile Carboys, Foxx Life Sciences
Bottles Carboys
EZBio® single use carboys are available in amber HDPE, dark amber polypropylene, HDPE, polyethylene terephthalate glycol-modified (PETG), and polycarbonate (PC).
  • Available in light blocking amber or clear formats
  • VersaCap® included
  • Gamma irradiated sterile
  • Double bag packaging
  • Polypropylene and polycarbonate carboys are autoclavable
  • PETG carboys are BPA-free
The VersaCap® technology and configurations increases the utility of these carboys for single-use applications. Made with USP Class VI components and gamma irradiated in the U.S., these carboysMore Product Information