Dynamic Climate Chambers, MKT 240, Binder
Supplier: Binder
9020-0363 9020-0364 9020-0365
76413-560EA 45680.7 USD
76413-560 76413-562 76413-564
Dynamic Climate Chambers, MKT 240, Binder
Testing Chambers Climatic Chambers
Standard climate chambers that work with APT.line™ preheating chamber technology.
  • Programmable condensation protection for test material
  • Heated viewing window with LED interior lighting
  • With 4 zero-voltage relay contacts
  • Troubleshooting system with visual and audible alarms
  • Access port with silicone plug (50 mm) at left
  • Computer interface: Ethernet
  • Power socket (230 V) on the right-side control panel
Intuitive touchscreen controller with time-segment and real-time programming. Internal data logger, measured values can be read out in open format viaMore Product Information