Vivaflow® 50R Ultrafiltration Units, Sartorius
Supplier: Sartorius
VF05H0 VF05H1 VF05H2 VF05H4
76408-654EA 354.04 USD
76408-654 76408-656 76408-658 76408-660
Vivaflow® 50R Ultrafiltration Units, Sartorius
Filters Centrifugal Filters
These are for 0.1 to 1000 ml, Hydrosart® membrane. Vivaflow 50R is a ready to use and reusable laboratory crossflow cassette for applications like cell culture supernatant concentration, virus concentration and water sample concentration. The thin channel, flip flow recirculation path of the modules provides high crossflow velocities with minimum pump speed requirements. A single module with 50 cm² active membrane area typically concentrates 250 ml by a factor of 20 in less than 30 min.
  • Fast and easy protein sample concentration
  • Reusable and optimal for
More Product Information