VWR® Advanced Cut Protection Gloves
Supplier: VWR International
76408-532 76408-534 76408-538 76408-540 76408-542
76408-532CS 967.33 USD
76408-532 76408-534 76408-538 76408-540 76408-542
VWR® Advanced Cut Protection Gloves
Gloves
The VWR® advanced cut protection glove with polyurethane coating provides enhanced cut protection for a range of applications that would include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coated
  • Gray knit glove with black coating
Ideal for use in environments handling items or tools with a high risk of cuts, including handling of items with sharp or jagged edges, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A3 Cut Protection (Level C). EN388:2016. The CEMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Latex/Latex-free Latex-Free
Powdered/Powder-free Powder-Free
Finish Polyurethane coating
Gauge 13
Sterility Non sterile