Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination with Controlled Auto Defrost Freezer, Horizon Scientific
Supplier: Horizon Scientific
PRF072WWG/0CAD PRF122WWW/0CAD PRF122WWG/0CAD
76407-438EA 5781.68 USD
76407-438 76407-440 76407-442
Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination with Controlled Auto Defrost Freezer, Horizon Scientific
Refrigerators
These White Diamond combination units provide both refrigeration and freezing capabilities in one convenient unit. These models are designed for scientific and clinical applications.
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Forced draft air circulation
  • Probe access
  • Keyed door locks
  • Controlled Auto Defrost technology
  • Natural hydrocarbon refrigerants
These combination units come equipped with forced draft circulation, and precise temperature monitoring through microprocessor control. The freezers on these units feature ControlledMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Cat. No. 76407-438 76407-440 76407-442
Defrost Controlled auto defrost freezer/cycle defrost refrigerator Controlled auto defrost freezer/cycle defrost refrigerator Controlled auto defrost freezer/cycle defrost refrigerator
Shelves/drawers 3 adjustable shelves (refrigerator), 2 adjustable shelves (freezer) 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer) 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
Door Type 1 glass swing door (refrigerator), 1 solid door (freezer) 2 solid swing doors 1 glass swing door (refrigerator), 1 solid door (freezer)
Shipping Weight 94 kg (207 lbs.) 143 kg (316 lbs.) 131 kg (289 lbs.)