Tubing for MINIPULS® 3 Peristaltic Pumps, Gilson®
Supplier: GILSON, INC.
F117740 F117742 F117744 F117746 F117747 F117748 F117749 F117750 F117933 F117934 F117935 F117936 F117938 F117939 F117940 F117941 F117942 F117943 F117945 F117946 F117948 F117949 F117952 F117953 F117954 F117956 F117958 F117960 F117962 F117963 F117965 F117966 F117968 F117969 F117970 F117975 F117976 F117977 F117978 F117979 F117980 F117981 F117982 F1825101 F1825102 F1825103 F1825111 F1825112 F1825113 F1825114 F1825122 F1825131 F1825132 F1825133 47526063 47526076 47526152 47526228 47526279 F1817741 F1817743 F1817745
76406-978EA 116.84 USD
76406-978 76406-980 76406-982 76406-984 76406-986 76406-988 76406-990 76406-992 76406-994 76406-996 76406-998 76407-000 76407-002 76407-004 76407-006 76407-008 76407-010 76407-012 76407-014 76407-016 76407-018 76407-020 76407-022 76407-024 76407-026 76407-028 76407-030 76407-032 76407-034 76407-036 76407-038 76407-040 76407-042 76407-044 76407-046 76407-048 76407-050 76407-052 76407-054 76407-056 76407-058 76407-060 76407-062 76407-064 76407-066 76407-068 76407-070 76407-072 76407-074 76407-076 76407-078 76407-080 76407-082 76407-084 76407-086 76407-088 76407-090 76407-092 76407-094 76407-096 76407-098 76407-100
Tubing for MINIPULS® 3 Peristaltic Pumps, Gilson®
Tubing Transfer Tubing
A variety of tubing of different material, inner diameter sizes, and lengths are available for MINIPULS® 3 pumps.
Silicone (Versilic®) tubing is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
ISOVERSINIC® tubing shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
PharMed® tubing meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
PVC (Tygon®) tubing handlesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable