High Performance Micro Balances, METTLER TOLEDO
Supplier: Mettler Toledo
30355547
76404-728EA 67098 USD
76404-728
High Performance Micro Balances, METTLER TOLEDO
Balances and Scales
With XPR micro balances we can able to weigh very small sample quantities directly into larger tare containers, with unrivalled accuracy up to 200 g.
  • 7" TFT color touchscreen (glove compatible)
  • The extremely high accuracy and low minimum weight make it possible to weigh very small quantities, leading to considerable material and cost savings
  • XPR micro-analytical balances have a readability range between 0.001 and 0.01 mg, and capacities up to 52 g, to cover all requirements for micro-analytical weighing
  • Ensure compliance with built-in functions
  • Multiple
More Product Information