VWR® HF Bottle Top Dispensers
Supplier: VWR International
76382-716 76382-718
76382-716EA 788.95 USD
76382-716 76382-718
VWR® HF Bottle Top Dispensers
Bottle Top Dispensers
Designed with ergonomics and intuitive handling in mind, HF bottle top dispenser helps in dispensing Hydrofluoric Acid with safety and precision.
  • Springless Valve™ for smooth and jam-free functioning
  • PTFE piston with ETP O-Ring for effortless piston movement and high chemical compatibility
  • Easy and effortless volume setting
  • Flexible delivery nozzle to change angle of dispensing
  • Dispensers is fully autoclavable at 121 °C and 15 psi for a duration of 10 to 15 minutes
  • Reproducibility of 0.2%
HF bottle top dispenser is designed with selected materials forMore Product Information