VWR® T Balances with Calibration Certificate
Supplier: VWR International
76378-438 76378-460 76378-462 76378-464 76378-466 76378-468 76378-470 76378-472 76458-486 76458-488 76458-490 76458-492 76458-494 76458-496 76458-498 76458-500
76378-438EA 6888.83 USD
76378-438 76378-460 76378-462 76378-464 76378-466 76378-468 76378-470 76378-472 76458-486 76458-488 76458-490 76458-492 76458-494 76458-496 76458-498 76458-500
VWR® T Balances with Calibration Certificate
Balances and Scales
The VWR® T-Series balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • 2×RS232, USB (A and B), Ethernet and Wi-Fi communication interfaces
  • Internal calibration
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
The VWR® T-Series balances with calibration certificate include an ISO 17025:2017 certificate of calibration showing the full testing data. TheseMore Product Information