Cubis® II Advanced Analytical Laboratory Balances, MCA Series, Standard Version, Sartorius
Supplier: Sartorius
MCA124S-2S00-U MCA324P-2S00-U MCA224S-2S00-U MCA524P-2S00-U MCA324S-2S00-U MCA124SU-S00 MCA124S-2S00-A MCA324P-2S00-A MCA124S-2S00-I MCA224S-2S00-A MCA524S-2S00-U MCA524P-2S00-A MCA324S-2S00-A MCA324P-2S00-I MCA224S-2S00-I MCA524P-2S00-I MCA324S-2S00-I MCA124SI-S00 MCA524SU-S00 MCA524S-2S00-A MCA224S-I-MTS MCA524S-2S00-I MCA324SI-S00 MCA524S-I-MTS
76376-962EA 8671.28 USD
76376-962 76377-006 76377-014 76377-056 76377-074 76377-082 76377-086 76377-150 76377-154 76377-156 76377-166 76377-178 76377-204 76377-208 76377-210 76377-222 76377-228 76377-254 76377-258 76377-264 76377-266 76377-280 76377-314 76377-318 76377-328 76377-346
Cubis® II Advanced Analytical Laboratory Balances, MCA Series, Standard Version, Sartorius
Balances and Scales
These Cubis® II laboratory balances are ideal for high-performance portfolio of both lab weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
  • Automatic glass draft shield with or without integrated ionizer to eliminate interfering electrostatic charges on samples and sample containers
  • Removable flat stainless-
More Product Information