Sodium chloride 0,9% USP Injection

Supplier: B BRAUN MEDICAL

Synonyms: Saline, NaCl, Sea salt, Halite

S8004-5384
76363-598CS 577.63 USD
76363-598
Sodium chloride 0,9% USP Injection
Sodium chloride

Sodium Chloride Injections USP are sterile, nonpyrogenic, isotonic and contain no bacteriostatic or antimicrobial agents. These intravenous solutions are indicated for use in adults and pediatric patients as sources of electrolytes and water for hydration.


Not manufactured with latex, PVC or DEHP.


NDC number: 00264-1800-31


Shelf life: 15 months (total)

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR