HiScale Columns, Cytiva
Supplier: Cytiva
28-9644-41 28-9644-44 28-9644-24 28-9644-45 28-9645-13 28-9645-14 29360550
10000-048EA 1365.9 USD
10000-048 10000-052 10000-054 10000-056 10000-058 10000-060 76358-650
HiScale Columns, Cytiva
Chromatography Columns
Empty columns with high pressure stability (able to withstand up to 20 bar of pressure) and high flow rates are compatible for packing with modern BioProcess™ chromatography media, such as MabSelect™ and Capto™ for process development and preparative chromatography. The columns are designed to be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of biomolecules, with the following few exceptions: chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol. Solutions containing more than 2 M NaOH, 1 M HCl, other strongMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. Pressure 20 bar
Temperature Limit 4...40 °C