CAPS (3-(cyclohexylamino)propanesulfonic acid)

Supplier: RPI

Synonyms: 3-Cyclohexylaminopropanesulfonic acid, 3-Cyclohexylaminopropanesulphonic acid

C30060-100.0 C30060-1000.0 C30060-500.0 C30060-250.0
76345-374EA 49.69 USD
76345-374 76345-376 76345-378 77510-204
CAPS (3-(cyclohexylamino)propanesulfonic acid)
CAPS (3-(Cyclohexylamino)propanesulphonic acid)

Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₆H₁₁NH(CH₂)₃SO₃H
Melting Pt: 300 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00003837
CAS Number: 1135-40-6

Order Now

Specification Test Results

Appearance White powder
Assay ≥99.0 %
Identification Pass
Loss on Drying (3HRS at 105 °C) ≤0.5 %
pH (1% in H2O) 4.9 - 6.0
PKa at 25 °C 10.3 - 10.5
Solubility Clear and Complete
UV Absorbance 260 nm (1 mol/L, 1 cm) ≤0.04
UV Absorbance 280 nm (1 mol/L, 1 cm) ≤0.02
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR