Micro IV Single-gas Detectors, GfG Instrumentation
Supplier: GFG INSTRUMENTATION, INC.
1418-022 1418-022K 1418-024 1418-024K 1418-027 1418-027K 1418-111 1418-111K 1418-114 1418-114K 1418-116 1418-116K 1418-120 1418-120K 1418-121 1418-121K 1418-126 1418-126K 1418-153 1418-153K 1418-170 1418-170K
76334-622EA 532.42 USD
76334-622 76334-624 76334-648 76334-650 76334-652 76334-654 76334-656 76334-658 76334-660 76334-662 76334-664 76334-666 76334-668 76334-670 76334-672 76334-674 76334-676 76334-678 76334-680 76334-682 76334-684 76334-686 76334-688 76334-690 76334-692
Micro IV Single-gas Detectors, GfG Instrumentation
Gas Detection Systems
The Micro IV single-gas detector is compact, robust and very easy to use. It is designed to be used in the harshest environments.
  • Highly configurable smart sensor design
  • Full 3-year warranty on O₂, CO and H₂S sensors
  • Event/data logging standard
  • Three programmable ceiling, STEL and TWA alarms
  • AutoCal® automatic calibration
It includes a set of rubber boots to withstand hard knocks and drops.
It keeps gas detection simple. Most day-to-day functions are controlled with a single button, and calibration is fully automatic and delivers up to 3000 hoursMore Product Information