One-Piece PTFE Lower Agitator, 13", 4-Blade, Chemglass
Supplier: Chemglass
CG-2095-M-55
76333-720EA 1773.53 USD
76333-720
One-Piece PTFE Lower Agitator, 13", 4-Blade, Chemglass
Stirrers Stirring Shafts
Agitator, one-piece, PTFE, four-blade, 30 mm, for 400 mm internal diameter flange 100 L reactor.
New and improved high viscosity agitator, the body is made entirely from ONE PIECE of CLASS VI VIRGIN PTFE. One-piece machined body is much easier to clean. The smooth machined surface has no seams, joints or crevices for material to collect. The design provides excellent stirring at the bottom of CG-1968-W 400 mm ID 100 L reactor vessels, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a PTFE safety pin. Heavy duty blades 9 mmMore Product Information