PIPETMAN® M Connected Electronic Pipettors, Gilson®
Supplier: GILSON, INC.
F81044 F81043 F81054 F81055 F81056 F81057 F81058 F81059 F81046 F81047 F81048 F81049 F81050 F81051 F81045 F81052 F81053 F81040 F81041 F81042
76333-064EA 1227.45 USD
76333-064 76333-066 76333-070 76333-072 76333-074 76333-076 76333-078 76333-080 76333-082 76333-084 76333-086 76333-088 76333-090 76333-092 76333-094 76333-096 76333-098 76409-312 76409-314 76409-316
PIPETMAN® M Connected Electronic Pipettors, Gilson®
Pipettes Micropipettes
PIPETMAN® M Connected is a Bluetooth®-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes. Requiring virtually no pipetting forces to aspirate and dispense samples, PIPETMAN M Connected helps reduce pipetting fatigue and increase pipetting efficiency. The pipette interacts in real-time with TRACKMAN® Connected making planning and executing pipetting protocols faster, easier, and more reliable.
  • Simple and intuitive motorized pipette
  • Ultimate consistency
  • Connect to Gilson Apps
With PipetteScope®,More Product Information