Memmert Paraffin Ovens, UNpa Series
Supplier: Memmert
UN75PA115VOLT UN110PA115VOLT UN160PA115VOLT UN55PA115VOLT UN30PA115VOLT
76332-886EA 5587.94 USD
76332-886 76332-888 76332-890 76332-936 76332-938
Memmert Paraffin Ovens, UNpa Series
Ovens Standard Ovens
Stainless steel interior, complete with manufacturers calibration certificate at 80 °C (measured at the center point of the chamber).
  • Stainless steel interior
  • Textured stainless steel exterior
  • Nearly gas tight chamber - no feed or exhaust air
  • Avoids danger of ignition of residue or damage to mechanical and electronic components
Reliable sample preparation for forensics, histology and microbiology. Five model sizes to chose from, sizes 32 to 161 litres volume. Twin Display units with high degree of thermal/heating safety with muliple overtemeratureMore Product Information