Avanti™ J-15 Series Benchtop Centrifuges, 200-230 V, Beckman Coulter®
Supplier: Beckman Coulter
C19389 C19390 C19395 C19396 C19401 C19402 C19407 C19408 C19413 C19414 C19419 C19420 C31546 C31547 C38175 C38176 C38177 C38178 C38179 C38180 C38181 C38182 C38183 C38184 C38185 C38186
76102-906EA 12308.5 USD
76102-906 76102-908 BKC19389 BKC19390 BKC19395 BKC19396 BKC19401 BKC19402 BKC19407 BKC19408 BKC19413 BKC19414 BKC19419 BKC19420 76332-088 76332-090 76332-092 76332-094 76332-096 76332-098 76332-100 76332-102 76332-104 76332-106 76332-108 76332-110 76332-112 76332-114
Avanti™ J-15 Series Benchtop Centrifuges, 200-230 V, Beckman Coulter®
Centrifuges Benchtop Centrifuges
The Avanti™ J-15 series of benchtop centrifuges (refrigerated or ventilated) leverage the ultra harmonic technology, designed to protect your sample and increase workflow time efficiencies.
  • 200-230 V models, not for use in USA
  • Ultra harmonic technology
  • Workflow optimization
  • Sample protection
  • Time efficiency
  • Multi-dimension imbalance tolerance
The Avanti™ J-15 series of benchtop centrifuges is part of a new family of life science equipment that provides the exceptional performance.
Refrigerated (Avanti™ J-15R) or ventilated (Avanti™ J-15), theseMore Product Information