Hypersil™ PREP BDS Si HPLC Columns
Supplier: Thermo Scientific
40305-259370A 40305-259570A 40310-059070A 40310-059270A 40310-109070A 40310-109270A 40310-159070A 40310-159270A 40310-159370A 40310-159570A 40310-259070A 40310-259270A 40310-259370A 40310-259570A 40310-018823A 40310-019023A 40310-019323A 40310-029323A 40305-059270XL 40305-109270XL 40305-159270XL 40305-159370XL 40305-159570XL 40305-259270XL 40305-259370XL 40305-259570XL 40310-059270XL 40310-109270XL 40310-159270XL 40310-159370XL 40310-159570XL 40310-259270XL 40310-259370XL 40310-259570XL 40303-052130 40303-102130 40303-152130 40303-252130 40305-052130 40305-102130 40305-152130 40305-252130 40303-054630 40303-104630 40303-154630 40303-254630 40305-054630 40305-104630 40305-154630 40305-254630 40305-059070A 40305-059270A 40305-109070A 40305-109270A 40305-159070A 40305-159270A 40305-159370A 40305-159570A 40305-259070A 40305-259270A
76330-842EA 8287.68 USD
76330-842 76330-844 76330-846 76330-848 76330-850 76330-852 76330-854 76330-856 76330-858 76330-860 76330-862 76330-864 76330-866 76330-868 76330-870 76330-872 76330-876 76330-878 76330-880 76330-882 76330-884 76330-886 76330-888 76330-890 76330-892 76330-894 76330-896 76330-898 76330-900 76330-902 76330-904 76330-906 76330-908 76330-910 76331-060 76331-062 76331-064 76331-066 76331-068 76331-070 76331-072 76331-074 76331-076 76331-078 76331-080 76331-082 76331-084 76331-086 76331-088 76331-090 76331-092 76331-094 76331-096 76331-098 76331-100 76331-102 76331-104 76331-106 76331-108 76331-110
Hypersil™ PREP BDS Si HPLC Columns
Chromatography Columns
Hypersil™ PREP BDS Si preparative HPLC columns which are packed with a base deactivated silica for normal phase chromatography of nonpolar and moderately polar organic compounds.
These preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Normal phase chromatography: Base deactivated silica to separate nonpolar and moderately polar organic compounds, USP L3, pH stable from 2 to 7, maximum temperature of 60 °C.
Standard threaded and standard flange hardware:
Designed forMore Product Information