Vesta-Syde® SQ 64 Ready-To-Use (RTU) Disinfectants
Supplier: Steris
634277 634296 634208
76311-092CS 455.26 USD
76311-092 76311-094 76311-096
Vesta-Syde® SQ 64 Ready-To-Use (RTU) Disinfectants
Cleaning Agents Disinfectants
STERIS' Vesta-Syde® SQ 64 ready to use quaternary ammonium disinfectant is specifically formulated for disinfection of hard surfaces. The ready to use solution can be applied manually or sprayed.
  • Ease of use - Eliminates the need to validate dilution preparation procedures by providing a consistent dilution every time
  • Convenience - Same key ingredients as Vesta-Syde® SQ disinfectant concentrate to simplify validation requirements
  • Productivity - Low residue formulation to maintain visibly clean surfaces with minimal rinsing
  • Efficacy - Microbial control
More Product Information