VWR® −40 °C Ultra-Low Temperature Freezers, 208- 230 V, Avantor
Supplier: VWR International
76307-986 76307-992 77661-484
76307-986EA 12905 USD
76307-986 76307-992 77661-484
VWR® −40 °C Ultra-Low Temperature Freezers, 208- 230 V, Avantor
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by green, hydrocarbon refrigerants delivering operational savings and sustainability.
  • 208-230 V models
  • Low energy
  • Low peak variation
  • Fast door opening recovery
  • Long warm up time
  • Three sizes
VWR -40 °C ULT freezers are built for seamless operation with user-friendly features. With low noise operation, flexible rack configurations and faster re-entry times, these -40 °C ULT freezers are designed to enhance the overall user experience and meet the needs of busy laboratories.
Certifications:
cULus, SNAP, CE
More Product Information