LyoTool™ Dual Plate Mop Adapter, Micronova
Supplier: Micronova
MZSM56-14 MZSM56-18 MZSM56-14IR MZSM56-18IR MZSM11-14 MZSM11-18 MZSM11-18IR MZSM11-14IR QDBP14-04 QDBP18-03 QDBP14-03
89131-858CS 632.82 USD
89131-858 89131-860 89131-864 89131-866 89236-640 89498-082 89498-084 10029-776 10040-504 76304-316 76304-360 76304-362
LyoTool™ Dual Plate Mop Adapter, Micronova
Mops
A dual-plated mop, specially designed to clean the top and bottom of lyophilizer shelves with one pass. The adjustable, compression fit plates ensure even pressure and consistent contact for optimal sanitization. Works well on any variety of equipment shelves spaced 3 to 6" apart.
  • Cleans top and bottom of shelves at the same time
  • Spring loaded plates for consistent pressure up and down
  • Front nose guard prevents extending beyond rack
  • Adjust to match shelf height with a simple twist of the knob
  • Use with SnapMop™ and PocketMop™ series flat head mop covers
More Product Information