Hi-T7 RNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0658S
76299-646EA 93 USD
76299-646
Hi-T7 RNA Polymerase, New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
Hi-T7 RNA Polymerase is an engineered DNA-dependent RNA polymerase that is highly specific for T7 phage promoters and is designed to function at higher temperatures than the wild-type bacteriophage T7 RNA Polymerase.
  • Active from 37 to 56 °C
  • For standard in vitro transcription at 37 °C use T7 RNA Polymerase
Hi-T7 RNA Polymerase functions at an optimal temperature of 50 to 52 °C.
Delivery information: Supplied with 10X Hi-T7 RNA Polymerase Reaction Buffer formulated for optimal performance.