Jacketed Filter Apparatus with 5000 ml Bottle, Ace Glass Incorporated
Supplier: Ace Glass
3710-02 3710-11 3710-12 3710-01
76291-830EA 3828.68 USD
76291-830 76291-832 76291-834 76291-894
Jacketed Filter Apparatus with 5000 ml Bottle, Ace Glass Incorporated
Funnels Filtration Funnels Filtration Apparatus Glass
Jacketed filtration apparatus or funnel for use with 75 mm membranes or filter paper. Ideal for solvent recycling and other filter applications where temperature control is required. Apparatus uses all threads to eliminate the need for clamping.
Complete apparatus includes a 5000 ml GL80 vacuum filtration wide mouth bottle, a jacketed filter funnel with 1" beaded pipe inlet/outlets, a PTFE Ace-Thred adapter, a Porosity C (25 to 50 micron) glass filter disc, and a PTFE retaining ring.
Funnel sold as separate piece is fully jacketed and has an Ace-Thred bottom and a beaded pipe inlet/More Product Information