Polynit Heatseal Wipes, Contec®
Supplier: Contec
PNHS-66 PN-99 IR LWPS0007 LWPS0006 PNHS-99B PNHS-99 PNHS-44 PNHS-99B/150 PNHS-1212 PNHS-1212B PNHS-22B LWPS0027 PNHS-44B PNHS-1620B
33501-474CS 1081.76 USD
33501-474 68310-183 68311-008 68311-010 62111-169 68300-556 68310-176 68310-180 68310-192 68310-194 89128-802 89217-354 10119-924 76289-764
Polynit Heatseal Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polynit Heatseal Wipes are made of 100% knitted polyester that is laser cut to bond the fibers at the edges of the wipe. This creates a very clean, non-abrasive edge that allows full utilization of the wipe surface.
  • Recommended for ISO Class 3–5 environments for nonsterile wipes
  • Recommended for Grade A/B or ISO Class 5 environments for sterile wipes
  • Laundered knitted 100% polyester fabric for extremely low levels of particles and fibers
  • Heat sealed edges help prevent particle contamination
  • 100% knitted polyester is abrasion and chemical resistant
  • Laser-cut,
More Product Information