Electronic Overhead Stirrers, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F201A0152 F201A0155 F201A0156 F201A0150 F201A0151 SA201A0151 SA201A0155
76288-116EA 1216.13 USD
76288-116 76288-118 76288-120 76288-172 76288-174 76288-176 76288-178
Electronic Overhead Stirrers, VELP Scientifica
Stirrers Overhead Stirrers
Complete range of overhead stirrers with electronic adjustment for a variety of needs in terms of viscosity and volume.
  • Wide range of applications: solubilization, mixing, emulsification and homogenization
  • Self-locking chuck simplifies assembly
  • Gentle start-up ensures optimum progression of the stirring speed
  • Most advanced safety standards
  • Through-hole to accept long shafts
Overhead stirrers with a technopolymer structure, ideal for premium resistance to acids, bases and solvents. Many reliable solutions are available, according to differentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Speed Range max 1300 rpm 50 - 2000 rpm, with SpeedServo™ to ensure constant speed and timer 50 - 2000 rpm 20 - 1200 rpm 50 - 2000 rpm, with SpeedServo™ to ensure constant speed and timer