Elite Series Waterproof pH Pocket Testers, Thermo Scientific
Supplier: Thermo Fisher Scientific
ELITEPH ELITEPHSPEAR
76284-650EA 150.31 USD
76284-650 76284-660
Elite Series Waterproof pH Pocket Testers, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Thermo Scientific™ Elite pH testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more.
  • Large back-lit graphic display
  • Uses 4 x AAA alkaline batteries
  • Average 150 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
Available with a direct-measure sensor or with a spear-shaped sensor.
One-year warranty for testers. Six-monthMore Product Information