Nalgene™ Economy Griffin Low-Form Plastic Beakers, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
1205-0050 1205-0100 1205-0150 1205-0250 1205-0400 1205-0600 1205-1000
76266-334CS 39.39 USD
76266-334 76266-336 76266-338 76266-408 76266-410 76266-412 76266-414
Nalgene™ Economy Griffin Low-Form Plastic Beakers, Polypropylene, Thermo Scientific
Beakers
Like the original Nalgene premium beaker, these are made from high quality laboratory grade polypropylene copolymer for reliably low extractables.
  • Molded-in graduations
  • Nest for compact storage
  • Autoclavable at 121 °C
  • Suitable for food contact
Molded-in graduations are of nominal accuracy and will not wear off over time. These beakers feature stable-footed base and "no-drip" pour spout for precision pouring.
Polypropylene copolymer is useful over a wide temperature range retaining its protective impact resistant properties to –40 °C. It isMore Product Information