Water Endotoxin_free, Sterile-Filtered

Supplier: QUALITY BIOLOGICAL, INC.

Synonyms: Dihydrogen oxide

118-325-101 118-325-131 118-325-101CS 118-325-131CS
76266-228EA 56.91 USD
76266-228 76266-230 76266-232 76266-234
Water Endotoxin_free, Sterile-Filtered
Water

Minimize contamination with Endotoxin-Free Water. Eliminate the risk of toxic activity produced by bacteria that can interfere with cell structure and function.


  • Mycoplasma tested
  • DNase, RNase and protease tested
  • Endotoxin < 0.005 EU/mL


Ideal for preparing solutions and reagents for transfection, transformation, cell and tissue cultures. Use when producing ultra-low, endotoxin serum-free media and buffers. Suitable when testing under pyrogen-free conditions (LAL testing) is required. Reliable for rinsing and washing equipment, glassware and plasticware in pyrogen-free conditions.

Sterile Filtered 0.1 µm
UNSPSC: 41116113
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 24 months


Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.


Delivery information: Shipping Condition: ambient


Caution: Research Use Only

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR