DURAN® GLS 80® Connection Cap System, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
293109103
76264-268CS 173.78 USD
76264-268
DURAN® GLS 80® Connection Cap System, DWK Life Sciences
Closures Bottle Closures
These GLS 80® screw caps with four ports and GL18 thread size quickly transfer multiple solutions.
  • Four ports
  • Choice of 5 inserts, each with different internal diameter depending on the application
  • Unused ports can be fitted with a blanking cap, if necessary
  • Sterile pressure compensation can be achieved using the membrane filter set (0.2 μm pore size)
  • The inserts and GL 18 screw connections hold the tubes securely in place, eliminating problems caused by slippage, as in HPLC plants
With easy-to-read scale and large labeling field for easy marking inMore Product Information