Plastic Policeman, United Scientific Supplies
Supplier: United Scientific Supplies
81411
76263-286PK 20.71 USD
76263-286
Plastic Policeman, United Scientific Supplies
Spatulas
Multi-purpose tool made from polypropylene can be used as a stirrer or spatula.
  • 9.5" length
  • Diameter of 0.25"
This dual purpose rod has a policeman on one end and a spatula on the other end.