Drying/Draining Rack, United Scientific Supplies
Supplier: United Scientific Supplies
81741
76263-280EA 184.41 USD
76263-280
Drying/Draining Rack, United Scientific Supplies
Racks Draining Racks
Large design is able to hold 20 pieces.
  • ABS plastic
  • 5" pegs
  • Waste tray included
The ABS plastic draining rack provides an excellent option for post-wash draining of tubes and bottles. The back plate has 20 holes into which pegs are tightened with the help of screw-nuts. The bottom of the rack features a waste water tray and drain.