End Mills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
206-001340 206-001239 206-001080 204-602826 204-001313 204-001200 204-001130 204-00110 204-001060 234-001080 206-001001 250-100150B 206-001040 229-001001 216-001001
76249-252EA 42.2 USD
76249-252 76249-254 76249-256 76249-258 76249-260 76249-262 76249-264 76249-266 76249-268 76250-410 76250-420 76250-422 76250-424 76250-928 76251-336
End Mills, Cutting Tool Supply
Tool Sets
End mills suitable for cast iron, non-ferrous metals, plastics and abrasive materials.
  • For profiling, slotting, production milling, finishing applications
  • Corner radius promotes additional strength for longer tool life resulting in greater productivity
  • Use for generating corner radius on work piece
  • Ball end allows for three axis contour slotting and contour finishing
Stub: reduces tool deflection and promotes rigidity - maintains excellent surface finishes
Regular: standard flute lengths for general all purpose milling
Long, X-long and extended lengths:More Product Information