Morse® Jobber Length Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
11684-MO 11683-MO 11676-MO 11675-MO 11657-MO 11656-MO 11655-MO 11654-MO 11653-MO 11652-MO 11650-MO 11649-MO 11648-MO 11644-MO 11643-MO 11640-MO 11639-MO 11635-MO 11634-MO 11633-MO 11632-MO 11629-MO 11628-MO 11627-MO 11626-MO 11625-MO 11623-MO 11619-MO 11617-MO 11616-MO 11615-MO 11613-MO 11608-MO 11603-MO 11598-MO 11593-MO 11590-MO 11589-MO 11586-MO 11580-MO 11577-MO 11576-MO 11575-MO 11574-MO 11573-MO 11404-MO 11384-MO
76249-150EA 26.59 USD
76249-150 76249-152 76249-154 76249-156 76249-158 76249-160 76249-162 76249-164 76249-166 76249-168 76249-170 76249-172 76249-174 76249-176 76249-178 76249-180 76249-182 76249-184 76249-186 76249-188 76249-190 76249-192 76249-194 76249-196 76249-198 76249-200 76249-202 76249-204 76249-206 76249-208 76249-210 76249-212 76249-214 76249-216 76249-218 76249-220 76249-222 76249-224 76249-226 76249-228 76249-230 76249-232 76249-234 76249-236 76249-238 76249-240 76249-242
Morse® Jobber Length Drills, Cutting Tool Supply
Tool Sets
These general-purpose straight shank jobber length drills are designed for drilling a wide variety of materials.
  • Made of high-speed steel
  • 118°-point style
  • Available in fractional, letter and wire gage sizes
  • Either of black oxide treated or bright finished drills
Black oxide surface treatment increases wear resistance, reduces galling and chip welding, improves chip flow and increases drill lubricant retention. Bright finish with polished flutes enhances chip ejection especially for aluminum and other non-ferrous materials.
Packaging: PackagingMore Product Information