FM-204 Self-Contained Desoldering and Soldering Station, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FM204-01
76248-054EA 2526.94 USD
76248-054
FM-204 Self-Contained Desoldering and Soldering Station, American Hakko Products
Soldering Supplies Soldering Stations
Self-contained desoldering and soldering station with a built-in vacuum pump. This versatile station uses the N3 series quick-change composite nozzles.
  • Vacuum pump type desoldering tool with excellent thermal recovery
  • Digital display ensures easy and reliable temperature control
  • Single-port station
  • Built in vacuum pump
  • Temperature accuracy ±15 °C
  • Gun or Pencil-style desoldering handpiece
  • Sleep mode functionality
Ordering information: Nozzles has to be ordered separately. Supplied with desoldering handpiece, filter pipe assembly, ceramic paper filter,More Product Information