Soldering Tips for C245 cartridge, JBC
Supplier: JBC TOOLS USA INC. TE
C245955 C245939 C245931 C245938 C245751 C245754 C245067 C245118 C245764 C245224 C245313 C245109 C245785 C245763 C245138 C245760 C245122 C245790 C245121
76240-746EA 259.41 USD
76240-746 76241-428 76241-430 76242-738 76244-166 76244-178 76245-542 76245-884 76247-496 76248-594 76248-832 76249-854 76249-856 76249-858 76249-860 76249-862 76250-140 76251-150 76251-210
Soldering Tips for C245 cartridge, JBC
Soldering Supplies Soldering Tips
C245 cartridge is the widest range with many shapes, sizes and special models.
Cartridge Cutter: Not tinned and designed for cutting.
Cartridge Knife: Knife tip with heating element.
Cartridge Drag: For through hole drag soldering.
Cartridge QFP: Designed for soldering QFP and PLCC.
Cartridge Fine Pitch: Used to reach joints on fine pitch high density connectors with more than two rows of extra long leads.
Cartridge for Plastics: Designed to be used in plastic applications.
Cartridge Pin/Connector: For soldering cables and connectors.
Cartridge Pin/More Product Information