Septihol® Ready-To-Use (RTU) Alcohol Solutions
Supplier: Steris
625408 625477 625405 625401
14223-886CS 388.4 USD
14223-886 14211-728 10736-414 76245-118
Septihol® Ready-To-Use (RTU) Alcohol Solutions
Cleaning Agents Disinfectants
STERIS' Septihol® Ready to use alcohol solution removes residue from hard surfaces. It can be used straight out of the container and be applied poured or sprayed. Since it contains no stabilizers or denaturants, it evaporates quickly and completely.
  • Easy to Validate - Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
  • Efficacy - Allows removal of process residues
  • Productivity - Quick evaporation with minimal residues
  • Ease of Use - Black trigger spray different color than sterile version to help identify the
More Product Information