Luer Lock Unassembled Manual Syringes, Jensen Global
Supplier: JENSEN GLOBAL INC
JGM5LL-20
76243-116PK 50.07 USD
76243-116
Luer Lock Unassembled Manual Syringes, Jensen Global
Syringes General-Purpose Syringes
These syringes come unassembled without any calibrations and are sold in packages with the barrel, stopper, and plunger.