Wrist Strap Monitor, SCS, Desco Industries
Supplier: Desco
725
76240-014EA 286.88 USD
76240-014
Wrist Strap Monitor, SCS, Desco Industries
Grounders Wrist Grounders
This wrist strap monitor visual and audible alarms indicate proper grounding of the operator and ground point.
Monitor can provide ground to a portable worksurface when connected through the 10 mm snap.
Certifications: Calibrated with NIST.