Dispensing Tips, OK International
Supplier: OK INTERNATIONAL TE
TT14-DHUV-PK TT18-DHUV-PK TT20-DHUV-PK TT22-DHUV-PK TT25-DHUV-PK TT16-DHUV-PK TT27-DHUV-PK
76239-416PK 90.58 USD
76239-416 76239-418 76239-420 76239-422 76239-424 76239-556 76240-408
Dispensing Tips, OK International
Syringe Needles
The tapered tip series designed with a double helix threads and universal luer lock to provide secure connection to industrial syringes or dispense valves.
  • Silicone and chloride-free with a maximum operating pressure 100 psi (6.9 bar)
  • Flash free orifice ensures accurate and repeatable dispensing
These tips are recommended for applications where dispensing medium and high-viscosity fluids with a high degree of precision and consistency is required. A smooth internal fluid path designed reduces turbulence flow, prevents air entrapment and fluid drippingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable