Oakton® Laboratory pH Electrode
Supplier: Antyila Scientific
35811-72
76221-312EA 197.04 USD
76221-312
Oakton® Laboratory pH Electrode
Electrodes pH Electrodes
Update, upgrade, or replace electrodes to get the best out of Oakton® meters.
  • All-in-one pH and temperature electrode
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.