N-Methyl-2-pyrrolidone (NMP), Low_water 99,0 GC_ASSAY_METHOD

Supplier: TCI America
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

M3055-500ML
76220-342EA 65.47 USD
76220-342
N-Methyl-2-pyrrolidone (NMP), Low_water 99,0 GC_ASSAY_METHOD
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Flash Pt: 95°C
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 14,06117

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Water max. 300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR