StaticAIRE™ Still Air Ionizer, Transforming Technologies
Supplier: Transforming Technologies
IN1000
76209-984EA 1462.49 USD
76209-984
StaticAIRE™ Still Air Ionizer, Transforming Technologies
Ionizers
The StaticAIRE™ Still Air Ionizer quickly and effectively eliminates static without using airflow. It’s designed for use in a variety of localized applications where space is limited and is ideal for installation inside equipment.
  • Quickly eliminate static charges in localized areas without airflow
  • Effective ionization range: 12"
  • Ideal where space is limited and installed inside equipment
  • Easy adjustment and calibration
An ideal product for mini-environments, analytical laboratories, and cleanrooms, the StaticAIRE™ Ionizer was designed to be virtuallyMore Product Information