CrossFLOW™ Wide Coverage Benchtop Ionizing Blower with Heater, Transforming Technologies
Supplier: Transforming Technologies
IN3000
76209-928EA 1581.78 USD
76209-928
CrossFLOW™ Wide Coverage Benchtop Ionizing Blower with Heater, Transforming Technologies
Ionizers
The CrossFLOW™ ionizing blower produces a powerful air flow, rich in positive and negative ions that quickly neutralize static charges.
  • Rapid static elimination performance
  • Wide coverage range: 3×6' foot area
  • Integrated heater
  • Integrated emitter point cleaner
Powered by inherently balanced AC technology, the ionizer provides a stable output with exceptionally fast decay performance. The CrossFLOW™ features built-in emitter point cleaners and a heater that can be turned on at any time for user comfort. It has an effective range of 3×6 ft. which canMore Product Information