ComfortTREAD™ Diamond-Plate Anti-Fatigue ESD Floor Mat, Transforming Technologies
Supplier: Transforming Technologies
FM9C FM9E
76209-908EA 234.85 USD
76209-908 76209-910
ComfortTREAD™ Diamond-Plate Anti-Fatigue ESD Floor Mat, Transforming Technologies
Floor Mats Traction and Anti-Fatigue Floor Mats
Quickly assemble an ESD floor that can handle heavy traffic, and can be installed by anyone.
  • Mats provide an anti-fatigue, energizing surface to stand on
  • Specially spaced diamond surface treads provide excellent traction
  • Available in interlocking sections to create runners in custom lengths
  • Wide beveled borders to promote safety
  • 100% conductive rubber
ComfortTREAD™ Diamond-Plate ESD Anti-Fatigue Mats keep workers comfortable and ESD sensitive equipment safe. The 100% conductive rubber mats are embossed with raised diamond-plate treads for superiorMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable