ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tubes, Simport Scientific
Supplier: Simport Scientific
T330-75NS T330-75AMS T330-76N T330-76B T330-76G T330-76R T330-76Y T330-76AM T330-75Y T330-75AM T330-76NS T330-76AMS T330-75N T330-75B T330-75G T330-75R T366NLS T366BLS T366GLS T366RLS T366WLS T366YLS T366AMLS T366NLSL T366BLSL T366GLSL T366RLSL T366WLSL T366YLSL T366AMLSL T365-5TN T365-5TAM T365-5AMLSS
76209-750CS 75.1 USD
76209-750 76209-752 76209-754 76209-756 76209-758 76209-760 76209-762 76209-764 76209-784 76209-786 76209-788 76209-790 76209-792 76209-794 76209-796 76209-798 76209-800 76209-802 76209-804 76209-806 76209-808 76209-810 76209-812 76209-814 76209-816 76209-818 76209-820 76209-822 76209-824 76209-826 76209-850 76209-852 76209-854 76209-856
ClikLok™ and Screw Cap MacrewTube™ Microcentrifuge Tubes, Simport Scientific
Tubes Microcentrifuge Tubes
The 5.0 ml ClikLok™ and MacrewTube™ microcentrifuge tubes have been designed for the simple and safe processing of medium-sized sample volumes.
  • Made of high-quality, transparent polypropylene without use of slip agents, plasticizers, mold release agents, and biocide substances
  • Centrifuge up to 25000 ×g
  • RNase, DNase, Pyrogen, and DNA-free
  • Operating temperature range from –196 to 121 °C
  • Autoclavable
  • Use the color-coded CapInsert™ for easy identification
Designed for samples larger than 2.0 ml. The 16mm tube diameter is identical to the diameter ofMore Product Information