Water DNAse_RNAse_free

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Dihydrogen oxide

04821932
76203-072EA 72.69 USD
76203-072
Water DNAse_RNAse_free
Water

DNase & RNase Free Water.


  • Key Applications : Investigation of nucleic acid and protein systems
  • Product Type : Lab Essentials
  • Lab Essential Type : Water
  • Chemical Class : Polar Protic
  • Refractive Index : n20/D 1.34(Lit.)
  • Presentation : Clear, Colorless Liquid
  • Format : Liquid
  • Foreign activity : DNase, RNase-Free
  • Sterility : 0.2 μm filtered
  • Certifications : DNase and RNase Free

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=637886a0f71965e5e696411e0a90ac128d618f6b&vid=b9180bed1b08d28a5d54c14ddbafa4824e7939e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR